accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1207-72-3

IUPAC Name: 10-methyl-10H-phenothiazine
Synonyms: 10H-phenothiazine, 10-methyl-
Molecular Weight (g/mol): 213.3
Molecular Formula: C13H11NS
InChi Key: QXBUYALKJGBACG-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2SC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)