accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12070-08-5

IUPAC Name: methanidylidynetitaniumylium
Synonyms: titanium carbide
Molecular Weight (g/mol): 59.88
Molecular Formula: CTi
InChi Key: YXIVWSJCLXKLJL-UHFFFAOYSA-N
SMILES: [C-]#[Ti+]
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)