accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12070-10-9

IUPAC Name: methanidylidynevanadiumylium
Synonyms: methanidylidynevanadiumylium
Molecular Weight (g/mol): 62.95
Molecular Formula: CV
InChi Key: ZLANVVMKMCTKMT-UHFFFAOYSA-N
SMILES: [C-]#[V+]
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)