accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NON

CAS RN 120747-84-4

IUPAC Name: 2-aminopyrimidine-5-carbaldehyde
Synonyms: 2-aminopyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 123.12
Molecular Formula: C5H5N3O
InChi Key: DPOYRRAYGKTRAU-UHFFFAOYSA-N
SMILES: NC1=NC=C(C=O)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results