accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONONSSOOHSOOH(E/Z)(E/Z)

CAS RN 1207638-53-6

IUPAC Name: 12-{5-[3-(carboxymethyl)-5-({4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,4H,8bH-cyclopenta[b]indol-7-yl}methylidene)-4-oxo-1,3-thiazolidin-2-ylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl}dodecanoic acid
Synonyms: 12-{5-[3-(carboxymethyl)-5-({4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,8bH-cyclopenta[b]indol-7-yl}methylidene)-4-oxo-1,3-thiazolidin-2-ylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl}dodecanoic acid
Molecular Weight (g/mol): 912.19
Molecular Formula: C52H53N3O6S3
InChi Key: HXSDFCKKOUCOIZ-UHFFFAOYNA-N
SMILES: OC(=O)CCCCCCCCCCCN1C(=S)SC(C1=O)=C1SC(=CC2=CC3=C(C=C2)N(C2CCCC32)C2=CC=C(C=C(C3=CC=CC=C3)C3=CC=CC=C3)C=C2)C(=O)N1CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)