accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 120800-50-2

IUPAC Name: 6-(piperidin-1-yl)pyridine-3-carboxylic acid
Synonyms: 6-(piperidin-1-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: QLPWWMSKVYYSEY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results