accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOFFFOO

CAS RN 1208222-39-2

IUPAC Name: 1-({[1,1'-biphenyl]-4-yl}methyl)-5-(trifluoromethoxy)-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 1-{[1,1'-biphenyl]-4-ylmethyl}-5-(trifluoromethoxy)indole-2,3-dione
Molecular Weight (g/mol): 397.35
Molecular Formula: C22H14F3NO3
InChi Key: SPBGRXOPAXZSER-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC2=C(C=C1)N(CC1=CC=C(C=C1)C1=CC=CC=C1)C(=O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results