accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClVCHCH

CAS RN 12083-48-6

IUPAC Name: bis(cyclopenta-2,4-dien-1-uid-1-yl)vanadiumbis(ylium) dichloride
Synonyms: bis(cyclopenta-2,4-dien-1-uid-1-yl)vanadiumbis(ylium) dichloride
Molecular Weight (g/mol): 252.03
Molecular Formula: C10H10Cl2V
InChi Key: UMGIRDUNLQIEBM-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[V++]([CH-]1C=CC=C1)[CH-]1C=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results