accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12084-29-6

IUPAC Name: barium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: barium acetylacetonate
Molecular Weight (g/mol): 335.55
Molecular Formula: C10H14BaO4
InChi Key: DJHZYHWLGNJISM-FDGPNNRMSA-L
SMILES: [Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results