CAS RN 12088-57-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis[tris(2,4-pentanedionato)titanium(IV)] Hexachlorotitanate(IV), TCI America™
CAS: 12088-57-2 Molecular Formula: C30H42Cl6O12Ti3 MDL Number: MFCD00059460 Synonym: Bis[tris(acetylacetonato)titanium(IV)] Hexachlorotitanate(IV)