accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1208985-45-8

IUPAC Name: 4,7,7-trimethyl-6-thiabicyclo[3.2.1]octane
Synonyms: trimethyl-6-thiabicyclooctane
Molecular Weight (g/mol): 170.31
Molecular Formula: C10H18S
InChi Key: FAXNZPOZWCWYBD-UHFFFAOYNA-N
SMILES: CC1CCC2CC1SC2(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results