accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12093-05-9

IUPAC Name: cycloocta-1,3,5,7-tetraene tris(methanidylidyneoxidanium) iron
Synonyms: 1,3,5,7-cyclooctatetraene tris(carbon monoxide) iron
Molecular Weight (g/mol): 244.03
Molecular Formula: C11H8FeO3
InChi Key: XAOCYVWRKJTXOL-UHFFFAOYSA-N
SMILES: [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1=CC=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results