accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHOOONOO

CAS RN 121-03-9

IUPAC Name: 2-methyl-5-nitrobenzene-1-sulfonic acid
Synonyms: 2-methyl-5-nitrobenzenesulfonic acid
Molecular Weight (g/mol): 217.2
Molecular Formula: C7H7NO5S
InChi Key: ZDTXQHVBLWYPHS-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1S(O)(=O)=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results