accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClFFF

CAS RN 121-17-5

IUPAC Name: 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-chloro-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: TZGFQIXRVUHDLE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)