CAS RN 121-27-7
CAS RN 121-27-7
IUPAC Name:
5-chloro-2-(4-chlorophenoxy)aniline
Synonyms:
5-chloro-2-(4-chlorophenoxy)aniline
Molecular Weight (g/mol):
254.11
Molecular Formula:
C12H9Cl2NO
InChi Key:
WLJSUJOESWTGEX-UHFFFAOYSA-N
SMILES:
NC1=CC(Cl)=CC=C1OC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
2-Amino-4,4'-dichlorodiphenyl Ether 98.0+%, TCI America™

CAS: 121-27-7 Molecular Formula: C12H9Cl2NO Molecular Weight (g/mol): 254.11 MDL Number: MFCD00027391 InChI Key: WLJSUJOESWTGEX-UHFFFAOYSA-N Synonym: 5-Chloro-2-(4-chlorophenoxy)aniline PubChem CID: 67135 IUPAC Name: 5-chloro-2-(4-chlorophenoxy)aniline SMILES: C1=CC(=CC=C1OC2=C(C=C(C=C2)Cl)N)Cl