accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-51-7

IUPAC Name: 3-nitrobenzene-1-sulfonyl chloride
Synonyms: 3-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 221.61
Molecular Formula: C6H4ClNO4S
InChi Key: MWWNNNAOGWPTQY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)