accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121-72-2

IUPAC Name: N,N,3-trimethylaniline
Synonyms: M-toluidine-N,N-dimethyl
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: CWOMTHDOJCARBY-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(C)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)