CAS RN 121-82-4
IUPAC Name:
1,3,5-trinitro-1,3,5-triazinane
Synonyms:
cyclotrimethylenetrinitramine hexogen RDX cyclonite
Molecular Weight (g/mol):
222.12
Molecular Formula:
C3H6N6O6
InChi Key:
XTFIVUDBNACUBN-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)N1CN(CN(C1)[N+]([O-])=O)[N+]([O-])=O
1
–
1
of
1
results
1
RDX, SPEX CertiPrep™
CAS: 121-82-4 Molecular Formula: C3H6N6O6 Molecular Weight (g/mol): 222.117 InChI Key: XTFIVUDBNACUBN-UHFFFAOYSA-N PubChem CID: 8490 ChEBI: CHEBI:24556 IUPAC Name: 1,3,5-trinitro-1,3,5-triazinane SMILES: C1N(CN(CN1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]