accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 1210-05-5

IUPAC Name: [1,1'-biphenyl]-2,2'-dicarbaldehyde
Synonyms: diphenaldehyde
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: HJFGULDHUDIPDA-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1C1=CC=CC=C1C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results