accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 1210-12-4

IUPAC Name: anthracene-9-carbonitrile
Synonyms: 9-anthracenecarbonitrile
Molecular Weight (g/mol): 203.24
Molecular Formula: C15H9N
InChi Key: KEQZHLAEKAVZLY-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CC=CC2=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results