CAS RN 1210-81-7
IUPAC Name:
[2-(5-chloro-2-methyl-1H-indol-3-yl)ethyl]dimethylamine; oxalic acid
Synonyms:
[2-(5-chloro-2-methyl-1H-indol-3-yl)ethyl]dimethylamine; oxalic acid
Molecular Weight (g/mol):
326.78
Molecular Formula:
C15H19ClN2O4
InChi Key:
FHSPBPPTDFRVGQ-UHFFFAOYSA-N
SMILES:
OC(=O)C(O)=O.CN(C)CCC1=C(C)NC2=CC=C(Cl)C=C12
1
–
1
of
1
results
1
ST 1936 oxalate, Tocris Bioscience™
CAS: 1210-81-7 Molecular Formula: C15H19ClN2O4 Molecular Weight (g/mol): 326.777 InChI Key: FHSPBPPTDFRVGQ-UHFFFAOYSA-N Synonym: st 1936 oxalate,5-chloro-3-2-dimethylamino ethyl-2-methylindole oxalate PubChem CID: 90488934 IUPAC Name: 2-(5-chloro-2-methyl-1H-indol-3-yl)-N,N-dimethylethanamine;oxalic acid SMILES: CC1=C(C2=C(N1)C=CC(=C2)Cl)CCN(C)C.C(=O)(C(=O)O)O