accessibility menu, dialog, popup

UserName

Viewing profile as user:
MnCH3CCOCOCO

CAS RN 12108-13-3

IUPAC Name: 1-methylcyclopenta-2,4-dien-1-ide tris(methanidylidyneoxidanium) manganese
Synonyms: methylcyclopentadienyl manganese tricarbonyl
Molecular Weight (g/mol): 218.09
Molecular Formula: C9H7MnO3
InChi Key: IYSGJCJSRBFZSZ-UHFFFAOYSA-N
SMILES: [Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C-]1C=CC=C1
Boiling Point 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)