CAS RN 121134-38-1
IUPAC Name:
3-(2-ethylhexyl)thiophene
Synonyms:
3-(2-ethylhexyl)thiophene
Molecular Weight (g/mol):
196.35
Molecular Formula:
C12H20S
InChi Key:
HWMQCIZYXLAJKM-UHFFFAOYNA-N
SMILES:
CCCCC(CC)CC1=CSC=C1
1
–
1
of
1
results
1
3-(2-Ethylhexyl)thiophene 97.0+%, TCI America™
CAS: 121134-38-1 Molecular Formula: C12H20S Molecular Weight (g/mol): 196.352 MDL Number: MFCD02260336 InChI Key: HWMQCIZYXLAJKM-UHFFFAOYSA-N PubChem CID: 14419543 IUPAC Name: 3-(2-ethylhexyl)thiophene SMILES: CCCCC(CC)CC1=CSC=C1