accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1211530-89-0

IUPAC Name: 2-bromo-3-fluoropyridine-4-carboxylic acid
Synonyms: 2-bromo-3-fluoropyridine-4-carboxylic acid
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: DHFIVEUMGCJILQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(Br)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results