accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClHNN

CAS RN 1212-48-2

IUPAC Name: hydrogen 2-benzyl-1H-1,3-benzodiazole chloride
Synonyms: hydrogen bendazole chloride
Molecular Weight (g/mol): 244.72
Molecular Formula: C14H13ClN2
InChi Key: CJTQARUHALKPGG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C(C1=NC2=CC=CC=C2N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)