accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFBr

CAS RN 121219-03-2

IUPAC Name: 4-bromo-3-fluorophenol
Synonyms: 4-bromo-3-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: MRQYTJXVULSNIS-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)