accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHFF

CAS RN 121219-16-7

IUPAC Name: (2,3-difluorophenyl)boronic acid
Synonyms: 2,3-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: SZYXKFKWFYUOGZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(F)=C1F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results