accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12125-08-5

IUPAC Name: diammonium hexachloroosmiumtetrakis(ylium)
Synonyms: diammonium hexachloroosmiumtetrakis(ylium)
Molecular Weight (g/mol): 439
Molecular Formula: Cl6H8N2Os
InChi Key: JRHMYOOMZKAPKT-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (4)
  • (1)
  • (2)
Grade 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (8)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)