accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeH3CH3CH3CCH3CH3H3CCH3H3CH3CCH3

CAS RN 12126-50-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 326.31
Molecular Formula: C20H30Fe
InChi Key: VKRKSGODPDKGAD-UHFFFAOYSA-N
SMILES: Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results