CAS RN 1213234-31-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Calcineurin Inhibitor VIII, CN585
The Calcineurin Inhibitor VIII, CN585, also referenced under CAS 1213234-31-1, controls the biological activity of Calcineurin. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.