accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSOH

CAS RN 121357-04-8

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)ethan-1-ol
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)ethanol
Molecular Weight (g/mol): 143.2
Molecular Formula: C6H9NOS
InChi Key: REKVZAFNJAMAQL-UHFFFAOYSA-N
SMILES: CC1=NC(CCO)=CS1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results