accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12136-58-2

IUPAC Name: dilithium(1+) sulfanediide
Synonyms: lithium sulfide
Molecular Weight (g/mol): 45.94
Molecular Formula: Li2S
InChi Key: GLNWILHOFOBOFD-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[S--]
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)