accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1214-47-7

IUPAC Name: (2E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
Synonyms: 2'-hydroxychalcone
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: AETKQQBRKSELEL-ZHACJKMWSA-N
SMILES: OC1=CC=CC=C1C(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results