accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12142-33-5

KKOSnOO

CAS RN 12142-33-5

IUPAC Name: dipotassium oxostannanebis(olate)
Synonyms: potassium stannate
Molecular Weight (g/mol): 244.9
Molecular Formula: K2O3Sn
InChi Key: IOUCSUBTZWXKTA-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Sn]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results