accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH2NFHCl

CAS RN 1214330-90-1

IUPAC Name: 1-(2-bromo-6-fluorophenyl)methanamine hydrochloride
Synonyms: 1-(2-bromo-6-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 240.5
Molecular Formula: C7H8BrClFN
InChi Key: WEGQVRTUTQKDNY-UHFFFAOYSA-N
SMILES: Cl.NCC1=C(F)C=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)