accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNNH3CSOOOH3CCH3CH3SOOOCH3FFNaNa

CAS RN 121461-69-6

IUPAC Name: disodium 2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide-5,11-disulfonate
Synonyms: disodium 2,2-difluoro-4,6,8,10,12-pentamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide-5,11-disulfonate
Molecular Weight (g/mol): 466.19
Molecular Formula: C14H15BF2N2Na2O6S2
InChi Key: FBAASMARNAYERO-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC1=C(C(C)=C2N1[B-](F)(F)[N+]1=C(C)C(=C(C)C1=C2C)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results