accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePP

CAS RN 12150-46-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 554.39
Molecular Formula: C34H28FeP2
InChi Key:
SMILES: [Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)