accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1215034-77-7

IUPAC Name: 2,3-difluoro-4-(trifluoromethyl)pyridine
Synonyms: 2,3-difluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 183.08
Molecular Formula: C6H2F5N
InChi Key: ZOMVJIOHXSXCIP-UHFFFAOYSA-N
SMILES: FC1=NC=CC(=C1F)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)