CAS RN 12152-94-2
IUPAC Name:
(cyclopenta-2,4-dien-1-yl)[2-(dihydroxyboranyl)cyclopenta-2,4-dien-1-yl]iron
Synonyms:
cyclopenta-2,4-dien-1-yl[2-(dihydroxyboranyl)cyclopenta-2,4-dien-1-yl]iron
Molecular Weight (g/mol):
229.85
Molecular Formula:
C10H11BFeO2
InChi Key:
HHGAJIKAUQWFKH-UHFFFAOYNA-N
SMILES:
OB(O)C1=CC=CC1[Fe]C1C=CC=C1
1
–
1
of
1
results
1
Ferroceneboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™
CAS: 12152-94-2 Molecular Formula: C10H11BFeO2 MDL Number: MFCD00059074