accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12158-57-5

IUPAC Name: titanium(4+) dicaesium(1+) trioxidandiide
Synonyms: caesium titanate
Molecular Weight (g/mol): 361.68
Molecular Formula: Cs2O3Ti
InChi Key: UKGZHELIUYCPTO-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Ti+4].[Cs+].[Cs+]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results