accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121591-98-8

IUPAC Name: 2-({6-[(4,5-dihydroxy-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-2-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxan-3-yl)oxy]-4,5-dihydroxy-2-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxan-3-yl}oxy)-6-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxane-3,4,5-triol
Synonyms: 2-({6-[(4,5-dihydroxy-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-2-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxan-3-yl)oxy]-4,5-dihydroxy-2-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxan-3-yl}oxy)-6-{[(3,4,5-trihydroxyoxan-2-yl)oxy]methyl}oxane-3,4,5-triol
Molecular Weight (g/mol): 1062.92
Molecular Formula: C39H66O33
InChi Key: PZUPAGRIHCRVKN-UHFFFAOYNA-N
SMILES: OCC1OC(O)C(O)C(O)C1OC1OC(COC2OCC(O)C(O)C2O)C(OC2OC(COC3OCC(O)C(O)C3O)C(OC3OC(COC4OCC(O)C(O)C4O)C(O)C(O)C3O)C(O)C2O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results