accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12168-54-6

IUPAC Name: oxo[(oxoferrio)oxy]iron; oxonickel
Synonyms: ferric oxide; nickel(II) oxide
Molecular Weight (g/mol): 234.38
Molecular Formula: Fe2NiO4
InChi Key: NQNBVCBUOCNRFZ-UHFFFAOYSA-N
SMILES: O=[Ni].O=[Fe]O[Fe]=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)