accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1216909-33-9

IUPAC Name: 2,7,10-trimethyl-9-(2,4,6-trimethylphenyl)acridin-10-ium perchlorate
Synonyms: 2,7,10-trimethyl-9-(2,4,6-trimethylphenyl)acridin-10-ium perchlorate
Molecular Weight (g/mol): 439.94
Molecular Formula: C25H26ClNO4
InChi Key: VDNXOHCMBBOSPE-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC1=CC2=C(C3=CC(C)=CC=C3[N+](C)=C2C=C1)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results