accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHONNHNH2OFFFHO

CAS RN 1217448-66-2

IUPAC Name: methyl 2-[2-(1-carbamimidoylpiperidin-3-yl)acetamido]-3-[4-(2-phenylethynyl)phenyl]propanoate; trifluoroacetic acid
Synonyms: methyl 2-[2-(1-carbamimidoylpiperidin-3-yl)acetamido]-3-[4-(2-phenylethynyl)phenyl]propanoate; trifluoroacetic acid
Molecular Weight (g/mol): 560.57
Molecular Formula: C28H31F3N4O5
InChi Key: BVMWPONRCOSTMK-UHFFFAOYNA-N
SMILES: OC(=O)C(F)(F)F.COC(=O)C(CC1=CC=C(C=C1)C#CC1=CC=CC=C1)NC(=O)CC1CCCN(C1)C(N)=N
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results