CAS RN 1217852-49-7
CAS RN 1217852-49-7
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylselanyl)butanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(methylselanyl)butanoic acid
Molecular Weight (g/mol):
418.36
Molecular Formula:
C20H21NO4Se
InChi Key:
MOIJGLAOGAWHTN-SFHVURJKSA-N
SMILES:
C[Se]CC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Selenomethionine, Advanced ChemTech
CAS: 1217852-49-7 Molecular Formula: C20H21NO4Se Molecular Weight (g/mol): 418.36 MDL Number: MFCD03093662 InChI Key: MOIJGLAOGAWHTN-SFHVURJKSA-N IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylselanyl)butanoic acid SMILES: C[Se]CC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O