CAS RN 1217863-33-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acetic acid-Trimethylpentane, 60/40 (v/v) soln., Thermo Scientific™
CAS: 1217863-33-6 MDL Number: MFCD00236632