accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12179-02-1

IUPAC Name: sodium hydrate hydroxide
Synonyms: sodium hydrate hydroxide
Molecular Weight (g/mol): 58.01
Molecular Formula: H3NaO2
InChi Key: JBJWASZNUJCEKT-UHFFFAOYSA-M
SMILES: O.[OH-].[Na+]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)