CAS RN 121808-62-6
IUPAC Name:
(4R)-3-[(2S)-5-oxopyrrolidine-2-carbonyl]-1,3-thiazolidine-4-carboxylic acid
Synonyms:
pidotimod
Molecular Weight (g/mol):
244.27
Molecular Formula:
C9H12N2O4S
InChi Key:
UUTKICFRNVKFRG-WDSKDSINSA-N
SMILES:
OC(=O)[C@@H]1CSCN1C(=O)[C@@H]1CCC(=O)N1