accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClMoHCHC

CAS RN 12184-22-4

IUPAC Name: molybdenum(4+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Synonyms: molybdocene dichloride
Molecular Weight (g/mol): 297.04
Molecular Formula: C10H10Cl2Mo
InChi Key: KJXKHGDHXCOQTK-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Mo+4].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results