accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121848-75-7

BrBr(?)

CAS RN 121848-75-7

IUPAC Name: 10,10'-dibromo-9,9'-bianthracene
Synonyms: 10,10'-dibromo-9,9'-bianthracene
Molecular Weight (g/mol): 512.24
Molecular Formula: C28H16Br2
InChi Key: NPNNLGXEAGTSRN-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=C(Br)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)